EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
1,25(OH)
PMID 28922720
Abstract
Breast cancer is one of the most common malignancies and bone is the commonest site of distant metastases. Evidences indicate that adequate supply of vitamin D will decrease the morbidity and mortality of breast cancer. However, the main role of vitamin D deficiency in breast cancer bone metastases remains unclear. In this study, the relationship between vitamin D and breast cancer bone metastases were evaluated. Results showed that 1,25(OH)