EMAIL THIS PAGE TO A FRIEND
The EMBO journal
Qualitative differences in T-cell activation by dendritic cell-derived extracellular vesicle subtypes.
PMID 28923825
Abstract
Exosomes, nano-sized secreted extracellular vesicles (EVs), are actively studied for their diagnostic and therapeutic potential. In particular, exosomes secreted by dendritic cells (DCs) have been shown to carry MHC-peptide complexes allowing efficient activation of T lymphocytes, thus displaying potential as promoters of adaptive immune responses. DCs also secrete other types of EVs of different size, subcellular origin and protein composition, whose immune capacities have not been yet compared to those of exosomes. Here, we show that large EVs (lEVs) released by human DCs are as efficient as small EVs (sEVs), including exosomes, to induce CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7888
DL-Propargylglycine, cystathionine γ-lyase inhibitor
C5H7NO2