EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Curcumin inhibited growth of human melanoma A375 cells via inciting oxidative stress.
PMID 28926928
Abstract
Curcumin, a polyphenol compound, possesses potent pharmacological properties in preventing cancers, which make it as a potential anti-cancer mediator. However, it is still unknown that whether Curcumin induced melanoma A375 cell was associated with oxidative stress. Here, we firstly found a fascinating result that Curcumin could reduce the proliferation and induced apoptosis of human melanoma A375 cells. Meanwhile, IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
70215
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence
C12H8O2