EMAIL THIS PAGE TO A FRIEND
Oncology letters
Clinicoradiological characteristics, management and prognosis of primary myeloid sarcoma of the central nervous system: A report of four cases.
PMID 28927153
Abstract
Myeloid sarcoma (MS) is a localized tumor composed of premature precursors of granulocytic cells, which may occur in any organ and most commonly involves the soft tissue and musculoskeletal system. This malignancy may occur in the presence or absence of hematological disorders. Primary MS involving the central nervous system (CNS-MS) is rare, and has only been described in a small number of isolated case reports. The diagnosis of CNS-MS is challenging and strategies for its management are undefined. The present study describes 4 cases of CNS-MS. The hematological indices at admission were normal and all patients presented with location-associated nonspecific symptoms. In magnetic resonance imaging scans, the tumors appeared isointense on T1-weighted and T2-weighted images, with marked enhancement following contrast agent administration. Only 1 patient progressed to acute myeloblastic leukemia (AML), with this occurring 3 weeks following histological diagnosis. During the follow-up period, the patient with AML succumbed to the disease, local recurrence was noted in another patient for which a second surgery was requested and no progression was observed in the remaining 2 patients. CNS-MS has unique radiological characteristics and, due to its diffuse tissue infiltration, gross total resection is challenging. It is important for clinicians to be aware of potential hematological disorders in patients with CNS-MS. A combined surgical and chemotherapeutic strategy may be able to provide long-term control of this malignancy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA035832
Anti-CD33 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA036722
Anti-CD34 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306481 ANTI-CD45(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA035304
Anti-CD99 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA056030
Anti-GFAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039179
Anti-LYG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA021147
Anti-MPO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004179
Anti-MUC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039835
Anti-NCAM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700750
Monoclonal Anti-CD117 antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
SAB4700772
Monoclonal Anti-Neurofilament medium protein antibody produced in mouse, clone NF-09, purified immunoglobulin, buffered aqueous solution