EMAIL THIS PAGE TO A FRIEND
Nature communications
Tumor-associated B-cells induce tumor heterogeneity and therapy resistance.
PMID 28928360
Abstract
In melanoma, therapies with inhibitors to oncogenic BRAF