EMAIL THIS PAGE TO A FRIEND
Frontiers in physiology
Aerobic Exercise Training Selectively Changes Oxysterol Levels and Metabolism Reducing Cholesterol Accumulation in the Aorta of Dyslipidemic Mice.
PMID 28928671
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
144657
Trifluoroacetamide, 97%
C2H2F3NO