EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
Molecular vaccine prepared by fusion of XCL1 to the multi-epitope protein of foot-and-mouth disease virus enhances the specific humoural immune response in cattle.
PMID 28929328
Abstract
Targeting antigen to dendritic cells (DCs) is a promising way to manipulate the immune response and to design prophylactic molecular vaccines. In this study, the cattle XCL1, ligand of XCR1, was fused to the type O foot-and-mouth disease virus (FMDV) multi-epitope protein (XCL-OB7) to create a molecular vaccine antigen, and an