EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
A lipophilic ionic liquid based on formamidinium cations and TFSI: the electric response and the effect of CO
PMID 28932830
Abstract
This work describes the preparation of the new lipophilic ionic liquid tetraoctyl-formamidinium bis(trifluoromethanesulfonyl) imide (TOFATFSI), which is miscible with lower alkanes. In particular, this work focuses on the electric behaviour of TOFATFSI in the particularly challenging highly apolar environment of supercritical CO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D201146
Dioctylamine, 97%
C16H35N
268607
Formamidine hydrochloride, 97%
CH4N2 · HCl