EMAIL THIS PAGE TO A FRIEND
Zebrafish
Deficiency of Serotonin in Raphe Neurons and Altered Behavioral Responses in Tryptophan Hydroxylase 2-Knockout Medaka (Oryzias latipes).
PMID 28933662
Abstract
Serotonin (5-hydroxytryptamine [5-HT]) is a bioactive monoamine that acts as a neurotransmitter in the central and peripheral nervous system of animals. Teleost fish species have serotonergic neurons in the raphe nuclei of the brainstem; however, the role of 5-HT in the raphe neurons in teleost fish remains largely unknown. Here, we established a medaka (Oryzias latipes) strain with targeted disruption of tryptophan hydroxylase 2 (tph2) gene that is involved in the 5-HT synthesis in the raphe nuclei. Immunohistochemistry and mass spectrometry analysis revealed that the homozygous mutants (tph2