PloS one
Purification of rabbit serum histidine-proline-rich glycoprotein via preparative gel electrophoresis and characterization of its glycosylation patterns.
PMID 28934288
Histidine-Proline-rich Glycoprotein (HPRG) is a plasma protein of vertebrates and several marine bivalves. Due to its multidomain structure consisting of several regions HPRG can interact with a variety of ligands, however the exact physiological role has not been discovered yet. Past purification approaches out of plasma or serum often led to co-purification of other proteins so that for a profound understanding of the function it is important to obtain a protein of high purity. Recent purification strategies were based upon metale chelate affinity chromatography followed by anion exchange chromatography or size exclusion chromatography, respectively. A large amount of serum albumin, the major plasma protein, also elutes from metale chelate affinity chromatography columns. Separation of rabbit HPRG from rabbit serum albumin could not be achieved via the above named methods by us. We present a method of purification of rabbit serum HPRG by means of metal affinity chromatography and preparative gel electrophoresis, which makes it possible to obtain HPRG practically devoid of impurities as assessed by mass spectrometry analysis. Moreover, we characterize the amount of glycosylation of HPRG and-to the best of our knowledge for the first time-the glycosylation pattern of rabbit HPRG.