EMAIL THIS PAGE TO A FRIEND
Chemosphere
Persulfate enhanced photocatalytic degradation of bisphenol A by g-C
PMID 28934651
Abstract
The enhancement of g-C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D23408
2,4-Diamino-6-phenyl-1,3,5-triazine, 97%
C9H9N5