EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Chemical characteristics and sources of PM
PMID 28935404
Abstract
During the 2016 Hangzhou G20 Summit, the chemical composition of submicron particles (PM