EMAIL THIS PAGE TO A FRIEND
Nature communications
Bioenergetic state regulates innate inflammatory responses through the transcriptional co-repressor CtBP.
PMID 28935892
Abstract
The innate inflammatory response contributes to secondary injury in brain trauma and other disorders. Metabolic factors such as caloric restriction, ketogenic diet, and hyperglycemia influence the inflammatory response, but how this occurs is unclear. Here, we show that glucose metabolism regulates pro-inflammatory NF-κB transcriptional activity through effects on the cytosolic NADH:NAD