EMAIL THIS PAGE TO A FRIEND
Frontiers in molecular neuroscience
RNA-Sequencing Analysis Reveals a Regulatory Role for Transcription Factor
PMID 28936162
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU028831 MISSION® esiRNA, esiRNA human FEZF2 (esiRNA1)