Progressive Fibrosis: A Progesterone- and KLF11-Mediated Sexually Dimorphic Female Response.
PMID 28938437
Progressive scarring is ubiquitous postoperatively and in an array of chronic systemic diseases. Recent studies indicate that such scarring has a high female propensity; females are also almost exclusively affected by endometriosis, a common sex steroid-dependent fibrotic disease. Endometriosis-related fibrosis is regulated epigenetically through transcription factor Krüppel-like factor 11 (KLF11). In response to surgical induction of endometriosis, Klf11-/- female mice develop significant fibrosis in contrast to wild-type mice. We therefore hypothesized that female fibrotic predilection was mediated by differential sex steroid regulation of KLF11/collagen 1a1 signaling and investigated the fibrotic response in wild-type and Klf11-/- male and female animals using a sterile peritonitis model. Fibrosis selectively developed in Klf11-/- females. Fibrosis in these animals was almost completely abrogated by ovariectomy. Ovariectomized animals were selectively supplemented with estradiol, medroxyprogesterone acetate (MPA), or dihydrotestosterone; fibrosis was only observed in mice exposed to MPA. Fibrosis therefore selectively developed in Klf11-/- female mice in response to physiological or pharmacological progesterone. The fibrotic response in these animals was also mitigated in response to antiprogestin therapy. Profibrotic gene expression was activated in a primary human peritoneal cell line in response to KLF11 short hairpin RNA and MPA but not estradiol. KLF11/collagen 1a1 signaling previously shown to be linked to fibrosis was thus selectively dysregulated in MPA-treated cells. Our in vivo and in vitro findings in an animal model and human cells, respectively, suggest that progressive fibrotic scarring is a sexually dimorphic response irrespective of etiology; moreover, it is responsive to novel, individualized therapeutic intervention.
Related Materials
Product #
Molecular Formula
Add to Cart
Chlorhexidine digluconate solution, 20% in H2O