EMAIL THIS PAGE TO A FRIEND
Oncotarget
PPA1 regulates tumor malignant potential and clinical outcome of colon adenocarcinoma through JNK pathways.
PMID 28938583
Abstract
Colorectal cancer (CRC) represents one of the most prevalent malignancies and the third leading cause of cancer death worldwide. Inorganic pyrophosphatase (PPA1) is an enzyme that catalyzes the hydrolysis of pyrophosphate to inorganic phosphate, therefore participates in the energy metabolism. Proteomic studies have demonstrated the up-regulated expression of PPA1 in various tumors, however, its expression pattern in CRC hasn't been reported. In the current study, we used RT-qCR, Western Blot and immunohistochemical (IHC) staining to explore the expression of PPA1 in 113 paired colon cancer tissues and adjacent normal tissues, which revealed that PPA1 was correlated with lymph node metastasis. The prognostic value of PPA1 was confirmed by Kaplan-Meier survival analysis and Cox regression analysis. We further purified PPA1 and obtained the phosphor-JNK1 protein and performed enzymatic studies, which identified that PPA1 can directly dephosphorylate pJNK1, while showed no catalytic activity towards pERK or p-p38 proteins. Moreover, overexpression of PPA1 enhanced cell viability through JNK-p53 signaling pathways, and it may also prevent cell apoptosis by inhibiting Bcl-2 and Caspase-3 cleavage. To our knowledge, this is the first study demonstrated the expression and clinical significance of PPA1 in colon cancer, which also provided evidence that figuring out PPA1 specific inhibitors can be invaluable in the future chemotherapy development towards colon cancer.