EMAIL THIS PAGE TO A FRIEND
Oncotarget
Calreticulin modulates the intracellular survival of mycobacteria by regulating ER-stress-mediated apoptosis.
PMID 28938588
Abstract
Endoplasmic reticulum (ER)-stress-mediated apoptosis is a host defense mechanism against