EMAIL THIS PAGE TO A FRIEND
Chinese journal of natural medicines
Terminalia arjuna bark extract attenuates picrotoxin-induced behavioral changes by activation of serotonergic, dopaminergic, GABAergic and antioxidant systems.
PMID 28939021
Abstract
Stress and emotion are associated with several illnesses from headaches to heart diseases and immune deficiencies to central nervous system. Terminalia arjuna has been referred as traditional Indian medicine for several ailments. The present study aimed to elucidate the effect of T. arjuna bark extract (TA) against picrotoxin-induced anxiety. Forty two male Balb/c mice were randomly divided into six experimental groups (n = 7): control, diazepam (1.5 mg·kg
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06244
Arjunic acid, analytical standard
C30H48O5
SMB00119
Arjunolic acid, ≥95% (LC/MS-ELSD)
C30H48O5