EMAIL THIS PAGE TO A FRIEND
The Journal of experimental biology
Microbiome symbionts and diet diversity incur costs on the immune system of insect larvae.
PMID 28939559
Abstract
Communities of symbiotic microorganisms that colonize the gastrointestinal tract play an important role in food digestion and protection against opportunistic microbes. Diet diversity increases the number of symbionts in the intestines, a benefit that is considered to impose no cost for the host organism. However, less is known about the possible immunological investments that hosts have to make in order to control the infections caused by symbiont populations that increase because of diet diversity. Using taxonomical composition analysis of the