Neurobiology of aging
A role of BAG3 in regulating SNCA/α-synuclein clearance via selective macroautophagy.
PMID 28941726
Many studies reveal that BAG3 plays a critical role in the regulation of protein degradation via macroautophagy. However, it remains unknown whether BAG3 affects the quality control of α-synuclein (SNCA), a Parkinson's disease-related protein. In this study, we demonstrated the increases of BAG3 expression in the ventral midbrain of SNCAA53T transgenic mice and also in MG132-treated PC12 cells overexpressing wild-type SNCA (SNCAWT-PC12). Moreover, we showed that BAG3 overexpression was sufficient to enhance the autophagy activity while knockdown of Bag3 reduced it in SNCAWT-PC12 cells. Immunoprecipitation revealed that BAG3 interacted with heat shock protein 70 and sequestosome 1. The immunostaining also showed the perinuclear accumulation and colocalization of BAG3 with these 2 proteins, as well as with LC3 dots in tyrosine hydroxylase-positive neurons in the midbrain of SNCAA53T mice. BAG3 overexpression was able to modulate SNCA degradation via macroautophagy which was prevented by Atg5 knockdown. Taken together, these results indicate that BAG3 plays a relevant role in regulating SNCA clearance via macroautophagy, and the heat shock protein 70-BAG3-sequestosome 1 complex may be involved in this process.