EMAIL THIS PAGE TO A FRIEND
Nature structural & molecular biology
Structural basis for specific cleavage of Lys6-linked polyubiquitin chains by USP30.
PMID 28945247
Abstract
Parkin ubiquitin (Ub) ligase (also known as PARK2) ubiquitinates damaged mitochondria for their clearance and quality control. USP30 deubiquitinase opposes parkin-mediated Ub-chain formation on mitochondria by preferentially cleaving Lys6-linked Ub chains. Here, we report the crystal structure of zebrafish USP30 in complex with a Lys6-linked diubiquitin (diUb or Ub