EMAIL THIS PAGE TO A FRIEND
Food chemistry
Dissipation rate study and pre-harvest intervals calculation of imidacloprid and oxamyl in exported Egyptian green beans and chili peppers after pestigation treatment.
PMID 28946222
Abstract
Two QuEChERS-based methods were developed and validated, using liquid chromatography-tandem mass spectrometric detection, in order to accurately determine residues of imidacloprid and oxamyl in green beans and chili peppers after treatment via irrigation system under field conditions in Egyptian farms. The validation included experiments for specificity, linearity, trueness, precision, matrix effect and limits of detection and quantification according to European Commission standards. The dissipation rates of both pesticides in green beans and chili peppers were studied and the pre-harvest intervals (PHIs) were calculated. The LOQ values of imidacloprid were 0.47 and 2.6μg/kg in green beans and chili peppers, respectively, while for oxamyl the LOQs were 2.9 and 0.67μg/kg, respectively. No PHI of imidacloprid is required, while for oxamyl it was found that still after 21days, its residues' concentration on both crops was significantly higher than the maximum residue limit.