EMAIL THIS PAGE TO A FRIEND
Cancer letters
HINT2 triggers mitochondrial Ca
PMID 28947137
Abstract
In early studies, it was shown that HINT2, which sensitizes cells to mitochondrial apoptosis, is down-regulated in hepatocellular carcinoma (HCC) cells (Martin et al., 2006). However, the molecular mechanism of this effect is unknown. Immunohistochemistry revealed that HINT2 expression is relatively low in pancreatic cancer tissues, compared to that in adjacent tissues (P < 0.05). Furthermore, its expression was related to pathological grade and lymph node metastasis (P = 0.0161 and 0.0108, respectively); in addition, down-regulation of HINT2 was found to be associated with relatively poor prognosis in pancreatic cancer patients. Up-regulation of HINT2 was shown to trigger pancreatic cancer cell apoptosis, decrease mitochondrial membrane potential (ΔΨm), promote intracellular reactive oxygen species (ROS) production, and elevate mitochondrial Ca