EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Kindlin supports platelet integrin αIIbβ3 activation by interacting with paxillin.
PMID 28954813
Abstract
Kindlins play an important role in supporting integrin activation by cooperating with talin; however, the mechanistic details remain unclear. Here, we show that kindlins interacted directly with paxillin and that this interaction could support integrin αIIbβ3 activation. An exposed loop in the N-terminal F
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0115
PAC-1, ≥98% (HPLC)
C23H28N4O2