EMAIL THIS PAGE TO A FRIEND
Frontiers in pharmacology
Ca
PMID 28955223
Abstract
Testosterone is known to induce cardiac hypertrophy through androgen receptor (AR)-dependent and -independent pathways, but the molecular underpinnings of the androgen action remain poorly understood. Previous work has shown that Ca
Related Materials