Clinical therapeutics
A Thorough QT/QTc Study of Clobazam in Healthy Volunteers.
PMID 28958437
A thorough QT study was conducted to assess the proarrhythmic potential of clobazam and its active metabolite, N-desmethylclobazam (N-CLB). In this Phase I, single-site, randomized, double-blind, double-dummy, parallel-group study, healthy participants were randomized to 1 of 4 treatment groups: clobazam 40 mg/d (maximum therapeutic dosage), clobazam 160 mg/d (supratherapeutic dosage), placebo, or moxifloxacin 400 mg (active control). Of 280 enrolled participants (n = 70 per treatment arm), 250 (92%) completed the study; 194 were included in the pharmacokinetics population (clobazam 40 mg/d, n = 66; clobazam 160 mg/d, n = 62; and moxifloxacin, n = 66). Mean changes from baseline in QT interval placebo-corrected for heart rate using the Fridericia formula (primary end point), Bazett formula, and individual correction method (QTcF, QTcB, and QTcI, respectively) with clobazam 40 and 160 mg/d revealed no effect on QTc. No clinically relevant or treatment-related arrhythmias were observed, and there were no instances of second- or third-degree atrioventricular block. Given that clobazam is primarily demethylated to N-CLB by cytochrome P450 (CYP) enzyme, CYP3A4, the mean plasma time-concentration profile of clobazam was unchanged with the exclusion of CYP2C19 poor metabolizers. As N-CLB is metabolized by CYP2C19, the exclusion of CYP2C19 poor metabolizers resulted in slightly decreased mean plasma time-concentration profiles of N-CLB. Using a linear mixed-effects model, the effects of the clobazam and N-CLB Cmax values on the placebo-corrected changes from baseline in QTcF, QTcI, and QTcB were near zero or slightly negative, and are not considered clinically important. The incidence of treatment-emergent adverse events was greatest in the clobazam groups (number of moderate AEs experienced by patients: PBO, 3/70; MOXI, 5/70; CLB 40 mg/d, 18/70; CLB 160 mg/d, 21/70; severe AEs: PBO, MOXI, & CLB 160 mg/d, 0; CLB 40 mg/d, 2/70); there were no serious AEs in any treatment group. A total of 10% of participants experienced benzodiazepine-withdrawal symptoms (16%, 23%, and 3% in the clobazam 40 and 160 mg/d groups and the moxifloxacin group, respectively). The findings from this thorough QT study are consistent with existing clinical data and support the lack of proarrhythmic potential with clobazam.
Related Materials
Product #
Molecular Formula
Add to Cart