EMAIL THIS PAGE TO A FRIEND
Food chemistry
Determination of parabens using two microextraction methods coupled with capillary liquid chromatography-UV detection.
PMID 28958548
Abstract
Parabens are common preservatives and environmental hormones. As such, possible detrimental health effects could be amplified through their widespread use in foods, cosmetics, and pharmaceutical products. Thus, the determination of parabens in such products is of particular importance. This study explored vortex-assisted dispersive liquid-liquid microextraction techniques based on the solidification of a floating organic drop (VA-DLLME-SFO) and salt-assisted cloud point extraction (SA-CPE) for paraben extraction. Microanalysis was performed using a capillary liquid chromatography-ultraviolet detection system. These techniques were modified successfully to determine four parabens in 19 commercial products. The regression equations of these parabens exhibited good linearity (r
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D221503
2-Dodecanol, 99%
C12H26O
X114
Triton X-114, laboratory grade