EMAIL THIS PAGE TO A FRIEND
Radiology and oncology
Effects of Electrochemotherapy with Cisplatin and Peritumoral IL-12 Gene Electrotransfer on Canine Mast Cell Tumors: a Histopathologic and Immunohistochemical Study.
PMID 28959165
Abstract
The study was aimed to characterize tumor response after combined treatment employing electrochemotherapy with IL-12 gene electrotransfer in dogs with spontaneous mast cell tumors (MCT). Eleven dogs with eleven MCT Histopathological examination of samples at T The combined electrochemotherapy and IL-12 gene electrotransfer effectively induced a cellular response against neoplastic cells characterized mainly by the recruitment of T-lymphocytes and macrophages and a fibrotic proliferation with reduction of microvessels.