EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Mutation in
PMID 28959249
Abstract
Many bacterial pathogens promote biofilms that confer resistance against stressful survival conditions. Likewise
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U4375
Uridine 5′-diphospho-N-acetylglucosamine sodium salt, ≥98%
C17H25N3Na2O17P2