EMAIL THIS PAGE TO A FRIEND
Frontiers in immunology
Systems Approach Reveals Nuclear Factor Erythroid 2-Related Factor 2/Protein Kinase R Crosstalk in Human Cutaneous Leishmaniasis.
PMID 28959260
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4441
DL-Sulforaphane, ≥90% (HPLC), synthetic, liquid
C6H11NOS2