EMAIL THIS PAGE TO A FRIEND
Protein science : a publication of the Protein Society
Web-accessible molecular modeling with Rosetta: The Rosetta Online Server that Includes Everyone (ROSIE).
PMID 28960691
Abstract
The Rosetta molecular modeling software package provides a large number of experimentally validated tools for modeling and designing proteins, nucleic acids, and other biopolymers, with new protocols being added continually. While freely available to academic users, external usage is limited by the need for expertise in the Unix command line environment. To make Rosetta protocols available to a wider audience, we previously created a web server called Rosetta Online Server that Includes Everyone (ROSIE), which provides a common environment for hosting web-accessible Rosetta protocols. Here we describe a simplification of the ROSIE protocol specification format, one that permits easier implementation of Rosetta protocols. Whereas the previous format required creating multiple separate files in different locations, the new format allows specification of the protocol in a single file. This new, simplified protocol specification has more than doubled the number of Rosetta protocols available under ROSIE. These new applications include pK
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS019977
Aldol, AldrichCPR
C4H8O2