EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Significance of hydrogen sulfide in sepsis-induced myocardial injury in rats.
PMID 28962136
Abstract
Sepsis-induced myocardial injury is a detrimental disorder for intensive care medicine due to its high rates of morbidity and mortality. Data suggest that nuclear factor (NF)-κB serves a critical role in the pathogenesis of myocardial injury. Hydrogen sulfide (H