EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Structural changes of waxy and normal maize starches modified by heat moisture treatment and their relationship with starch digestibility.
PMID 28962764
Abstract
In the present study, the variations in structure of waxy and normal maize starches modified by heat-moisture treatment (HMT) for different treating time (3h and 9h) were investigated. HMT caused the destruction of starch granules. The
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3176
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid