Carbohydrate polymers
Fabrication of cellulose acetate/polybenzoxazine cross-linked electrospun nanofibrous membrane for water treatment.
PMID 28962782
Herein, polybenzoxazine based cross-linked cellulose acetate nanofibrous membrane exhibiting enhanced thermal/mechanical properties and improved adsorption efficiency was successfully produced via electrospinning and thermal curing. Initially, suitable solution composition was determined by varying the amount of the benzoxazine (BA-a) resin, cellulose acetate (CA) and citric acid (CTR) to obtain uniform nanofibrous membrane via electrospinning. Subsequently, thermal curing was performed by step-wise at 150, 175, 200 and 225°C to obtain cross-linked composite nanofibrous membranes. SEM images and solubility experiments demonstrated that most favorable result was obtained from the 10% (w/v) CA, 5% (w/v) BA-a and 1% (w/v) CTR composition and cross-linked nanofibrous membrane (CA10/PolyBA-a5/CTR1) was obtained after the thermal curing. Chemical structural changes (ring opening) occurred by thermal curing revealed successful cross-linking of BA-a in the composite nanofibrous membrane. Thermal, mechanical and adsorption performance of pristine CA and CA10/PolyBA-a5/CTR1 nanofibrous membranes were studied. Char yield of the pristine CA nanofibrous membrane has increased notably from 12 to 24.7% for composite CA10/PolyBA-a5/CTR1 membrane. When compared to pristine CA membrane, CA10/PolyBA-a5/CTR1 nanofibrous membrane has shown superior mechanical properties having tensile strength and Young's modulus of 8.64±0.63MPa and 213.87±30.79MPa, respectively. Finally, adsorption performance of pristine CA and CA10/PolyBA-a5/CTR1 nanofibrous membranes was examined by a model polycyclic aromatic hydrocarbon (PAH) compound (i.e. phenanthrene) in aqueous solution, in which CA10/PolyBA-a5/CTR1 nanofibrous membrane has shown better removal efficiency (98.5%) and adsorption capacity (592μg/g).
Related Materials
Product #
Molecular Formula
Add to Cart
Phenanthrene, 98%