EMAIL THIS PAGE TO A FRIEND
Nature communications
Slow-wave sleep is controlled by a subset of nucleus accumbens core neurons in mice.
PMID 28963505
Abstract
Sleep control is ascribed to a two-process model, a widely accepted concept that posits homoeostatic drive and a circadian process as the major sleep-regulating factors. Cognitive and emotional factors also influence sleep-wake behaviour; however, the precise circuit mechanisms underlying their effects on sleep control are unknown. Previous studies suggest that adenosine has a role affecting behavioural arousal in the nucleus accumbens (NAc), a brain area critical for reinforcement and reward. Here, we show that chemogenetic or optogenetic activation of excitatory adenosine A
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2765
Ibotenic acid, ~95%, solid
C5H6N2O4