EMAIL THIS PAGE TO A FRIEND
Journal of molecular neuroscience : MN
Concentration-Dependent Inner Retina Layer Damage and Optic Nerve Degeneration in a NMDA Model.
PMID 28963708
Abstract
The intravitreal injection of N-methyl-D-aspartate (NMDA), a glutamate analogue, is an established model for fast retinal ganglion cell (RGC) degeneration. Yet, NMDA does not cause specific RGC damage. Now, the effects on the whole retina were analyzed. Additionally, the related effects for the structure and apoptotic levels of the optic nerve were investigated. Therefore, different NMDA concentrations were intravitreally injected in rats (20, 40, or 80 nmol NMDA or PBS). At days 3 and 14, Brn-3a