Journal of photochemistry and photobiology. B, Biology
The interaction between the light source dose and caspase-dependent and -independent apoptosis in human SK-MEL-3 skin cancer cells following photodynamic therapy with zinc phthalocyanine: A comparative study.
PMID 28964887
The aim of this study is to determine the behavior of relative expression of Bcl-2, caspase-8, caspase-9, and caspase-3 genes of/in SK-MEL-3 cancer cells and explore molecular mechanisms responsible for the apoptosis response during an in vitro photodynamic therapy (PDT) with Zinc Phthalocyanine (ZnPc) using different doses of the light source. In this study, firstly the cytotoxic effects of ZnPc-PDT on SK-MEL-3 cells were evaluated. By irradiating the laser, ZnPc induced a significant amount of apoptosis on SK-MEL-3 cells in three IC50s including 0.064±0.01, 0.043±0.01, and 0.036±0.01μg/mL at the doses of 8, 16, and 24J/cm2, respectively. Moreover, flow cytometry and QRT-PCR experiments were done. The high percentage of apoptotic cells was seen in the early apoptosis stage. The expression of Bcl-2 and caspase-8 genes at all doses of laser experienced an obvious reduction in comparison to the control group. On the other hand, although the expression of caspase-9 and caspase-3 genes remains almost constant at 8J/cm2, but they faced an increment at 16 and 24J/cm2 doses. These data reveal caspase-dependent apoptosis in high and caspase-independent apoptosis in low doses of laser. Based on the results of present work, it can be suggested that the dose of the light source is a key factor in induction of caspase-dependent and caspase-independent apoptosis pathways following PDT.
Related Materials
Product #
Molecular Formula
Add to Cart
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine, ≥99%