EMAIL THIS PAGE TO A FRIEND
Biophysical chemistry
Bioactive compounds from extra virgin olive oils: Correlation between phenolic content and oxidative stress cell protection.
PMID 28965785
Abstract
When compared with other edible vegetable oils, the extra virgin olive oil (EVOO) exhibits excellent nutritional properties due to the presence of biophenolic compounds. Although they constitute only a very small amount of the unsaponifiable fraction of EVOO, biophenols strongly contribute to the sensorial properties of this precious food conferring it, for example, the bitter or pungent taste. Furthermore, it has been found that biophenols possess beneficial effects against many human pathologies such as oxidative stress, inflammation, cardiovascular diseases, cancer and aging-related illness. In the present work, the biophenolic content of 51 Italian and Spanish EVOOs was qualitatively and quantitatively identified and their antioxidant ability analyzed by oxygen radical absorbance capacity (ORAC) assay. Results indicated that the maximum relationship can be found if the ORAC value is correlated with the concentration of the large family composed by ligstroside and oleuropein derivatives together with their degradation products, hydroxytyrosol and tyrosol. Then, selected biophenolic extracts were tested in NIH-3T3 cell line to verify their ability in the recovery of the oxidative stress revealed by DCFH-DA assay. Results were linearly correlated with the concentration of ligstroside aglycone (aldehyde and hydroxyl form).