EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Antimicrobial and Antibiofilm Potential of Acyclic Amines and Diamines against Multi-Drug Resistant
PMID 28966610
Abstract
Multi-drug resistant
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51387
MRSA Selective Supplement, for microbiology
C17H20N2O6S