EMAIL THIS PAGE TO A FRIEND
Oncotarget
Bufalin-loaded bovine serum albumin nanoparticles demonstrated improved anti-tumor activity against hepatocellular carcinoma: preparation, characterization, pharmacokinetics and tissue distribution.
PMID 28968991
Abstract
To prepare and evaluate the liver-targeted drug delivery system of Bufalin with higher liver uptake and stronger antitumor activity against hepatocellular carcinoma. Bufalin-loaded bovine serum albumin nanoparticle was prepared by desolvation method, to investigate the The Bufalin-loaded bovine serum albumin nanoparticle with an average particle size of 125.1 nm exhibited a sustained release behavior Bufalin-loaded bovine serum albumin nanoparticle is a promising liver-targeted drug delivery system with higher liver uptake and stronger antitumor activity against hepatocellular carcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0261
Bufalin
C24H34O4