EMAIL THIS PAGE TO A FRIEND
Oncotarget
Repression of intestinal transporters and FXR-FGF15 signaling explains bile acids dysregulation in experimental colitis-associated colon cancer.
PMID 28969019
Abstract
Bile acids (BAs) are important endogenous signaling molecules that play vital roles in the pathological development of various diseases including colitis-associated cancer (CAC). BAs were previously found dysregulated under conditions of CAC; however, the exact patterns and underlying molecular mechanisms remain largely elusive. Based on the development of a method for comprehensive analysis of BAs, this study aims to elucidate the dysregulation patterns and involved mechanisms in a typical CAC model induced by azoxymethane (AOM)/dextran sodium sulfate (DSS). CAC mice showed decreased BAs transformation in gut and glucuronidation in colon, leading to accumulation of primary BAs but reduction of secondary BAs in colon. CAC mice were characterized by an accumulation of BAs in various compartments except ileum, which is in line with repressed ileal FXR-FGF15 feedback signaling and the increased expression of hepatic CYP7A1. The compromised ileal FXR-FGF15 signaling was caused in part by the reduced absorption of FXR ligands including free and tauro-conjungated BAs due to the downregulation of various transporters of BAs in the ileum of CAC mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3878
Hyodeoxycholic acid, ≥98%
C24H40O4