EMAIL THIS PAGE TO A FRIEND
Oncotarget
Anti-proliferation of triple-negative breast cancer cells with physagulide P: ROS/JNK signaling pathway induces apoptosis and autophagic cell death.
PMID 28969050
Abstract
Physagulide P (PP), a new natural compound, was isolated from