EMAIL THIS PAGE TO A FRIEND
Molecular pharmaceutics
Targeted Cancer Therapy Using Fusion Protein of TNFα and Tumor-Associated Fibronectin-Specific Aptide.
PMID 28969419
Abstract
Tumor necrosis factor-α has shown potent antitumor effects in preclinical and clinical studies. However, severe side effects at less than therapeutic doses have limited its systemic delivery, prompting the need for a new strategy for targeted delivery of the protein to tumors. Here, we report a fusion protein of mouse tumor necrosis factor (TNF)-α (mTNFα) and a cancer-targeting, high-affinity aptide and investigate its therapeutic efficacy in tumor-bearing mice. A fusion protein consisting of mTNFα, a linker, and an aptide specific to extra domain B (EDB) of fibronectin (APT
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2011
Melphalan, powder
C13H18Cl2N2O2