EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Simultaneous adsorption of aflatoxin B
PMID 28972897
Abstract
Organo-montmorillonites (OMts) modified with mono- and di-alkyl cationic surfactants were prepared to remove polar mycotoxin aflatoxin B