EMAIL THIS PAGE TO A FRIEND
Oncotarget
Effects of liraglutide on hemodynamic parameters in patients with heart failure.
PMID 28977981
Abstract
Glucagon-like peptide-1 analogues improve left ventricular function in patients with acute myocardial infarction. This study aimed to evaluate the effects of liraglutide on hemodynamic parameters in patients with heart failure. A total of 78 patients with heart failure were enrolled in this study between August 2014 and November 2015. Of these, 52 patients were randomized 1:1 to receive either liraglutide or placebo for 7 days. Hemodynamic measurements were made using transpulmonary thermodilution and arterial pulse contour analysis. At 7 days, the difference in change of the primary endpoint of cardiac output between the liraglutide group and control group was +1.1 1/min (95% CI +0.1 to +2.2;