EMAIL THIS PAGE TO A FRIEND
Oncotarget
Vitamin D deficiency causes insulin resistance by provoking oxidative stress in hepatocytes.
PMID 28978056
Abstract
Vitamin D deficiency could cause insulin resistance. However, the underlying mechanisms are unclear. The 1α-Hydroxylase ["1α(OH)ase"] is a key enzyme for activate vitamin D3 synthesis. Here, we show that 1α(OH)ase stable knockdown by targeted shRNA led to vitamin D3 depletion in L02 hepatocytes. 1α(OH)ase silence also inhibited insulin-induced downstream signaling (IRS-1, ERK and AKT) transduction and glucose transporter 4 expression. Further, 1α(OH)ase shRNA in L02 hepatocytes led to significant reactive oxygen species production, p53-p21 activation and DNA damages. Such effects were almost completely reversed with co-treatment of n-acetylcysteine, which is an established anti-oxidant. Remarkably, insulin-induced downstream signaling transduction and glucose transporter 4 expression were recovered with n-acetylcysteine co-treatment in 1α(OH)ase-silenced L02 hepatocytes. Together, our results suggest that vitamin D deficiency-induced insulin resistance is possibly caused by oxidative stress in hepatocytes.