EMAIL THIS PAGE TO A FRIEND
Oncotarget
Tunicamycin induced endoplasmic reticulum stress promotes apoptosis of prostate cancer cells by activating mTORC1.
PMID 28978108
Abstract
Studies suggest that tunicamycin may work as a therapeutic drug to cancer cells by inducing stress in the endoplasmic reticulum (ER) through unfolded protein response (UPR) and thereby promoting apoptosis. However, mechanisms of the prolonged activation of the UPR under sustained ER stress in the regulation of cell apoptosis are largely unknown. To delineate the role of candidate genes in the apoptotic process under ER stress and to search for new therapeutic strategies to treat metastatic castration resistant prostate cancer, we performed whole genome expression microarray analysis in tunicamycin treated metastatic androgen-insensitive prostate cancer cells, PC-3. Among several induced genes, the expression of eNOS (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU027651 MISSION® esiRNA, esiRNA human SQSTM1 (esiRNA1)