EMAIL THIS PAGE TO A FRIEND
Iranian journal of parasitology
The Potential Use of Methotrexate in the Treatment of Cutaneous Leishmaniasis: In Vitro Assays against Sensitive and Meglumine Antimoniate-resistant Strains of
PMID 28979343
Abstract
The present study aimed to evaluate the effect of methotrexate (MTX) alone and in combination with meglumine antimoniate (MA, Glucantime) against sensitive and MA-resistant The present study was carried out in 2014 in Leishmaniasis Research Center at School of Medicine, Kerman University of Medical sciences, Kerman, Iran. The effects of MTX alone and along with MA on promastigote and amastigote stages of sensitive (SS) and MA-resistant (RS) The findings of OD and IC This study indicated high potency and a synergistic effect of MTX on MA in inhibiting the growth rateof promastigote and amastigote stages of sensitive and meglumine antimoniate-resistant