EMAIL THIS PAGE TO A FRIEND
3 Biotech
Bioconversion of isoeugenol to vanillin and vanillic acid using the resting cells of
PMID 28979831
Abstract
40 isoeugenol-tolerant yeasts were isolated from the rhizosphere soil samples which in turn were collected from aromatic plants in different regions of Iran, and further tested for their ability to grow on a minimal medium containing isoeugenol as the sole carbon and energy source. Nine isolates which were able to grow on isoeugenol were examined for their ability to convert isoeugenol into vanillin under growing cell experiments. Of the tested yeasts, the highest conversion efficiency was observed in isolate MP24. The isolate was identified as
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I17206
Isoeugenol, 98%, mixture of cis and trans
C10H12O2
V1104
Vanillin, ReagentPlus®, 99%
C8H8O3