American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics
Nhe5 deficiency enhances learning and memory via upregulating Bdnf/TrkB signaling in mice.
PMID 28981195
Nhe5, a Na+ /H+ exchanger, is predominantly expressed in brain tissue and is proposed to act as a negative regulator of dendritic spine growth. Up to now, its physiological function in vivo remains unclear. Here we show that Nhe5-deficient mice exhibit markedly enhanced learning and memory in Morris water maze, novel object recognition, and passive avoidance task. Meanwhile, the pre- and post-synaptic components, synaptophysin (Syn) and post-synaptic density 95 (PSD95) expression levels were found increased in hippocampal regions lacking of Nhe5, suggesting a possible alterations in neuronal synaptic structure and function in Nhe5-/- mice. Further study reveals that Nhe5 deficiency leads to higher Bdnf expression levels, followed by increased phosphorylated TrkB and PLCγ levels, indicating that Bdnf/TrkB signaling is activated due to Nhe5 deficiency. Moreover, the corresponding brain regions of Nhe5-/- mice display elevated ERK/CaMKII/CREB phosphorylation levels. Taken together, these findings uncover a novel physiological function of Nhe5 in regulating learning and memory, further implying Nhe5 as a potential therapeutic target for improving cognition.
Related Materials